Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
FGF14 is involved in nervous system development and function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA397O8.2; FGF; FGF homologus factor 4b; FGF-14; FHF-4; Fibroblast growth factor; Fibroblast growth factor 14; fibroblast growth factor 14, isoform b; Fibroblast growth factor homologous factor 4; m 1B
Gene Aliases: FGF-14; FGF14; FHF-4; FHF4; mFHF-4(1B); SCA27
UniProt ID: (Human) Q92915, (Rat) Q8R5L7, (Mouse) P70379
Entrez Gene ID: (Human) 2259, (Rat) 63851, (Mouse) 14169
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support