Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MYSAPSACT CLCLHFLLLC FQVQVLVAEE NVDFRIHVEN QTRARDDVSR KQLRLYQLYS RTSGKHIQVL GRRISARGED GDKYAQLLVE TDTFGSQVRI KGKETEFYLC MNRKGKLVGK PDGTSKECVF IEKVLENNYT ALMSAKYSGW YVGFTKKGRP RKGPKTRENQ QDVHFMKRYP KGQPELQKPF KYTTVTKRSR RIRPTHPA (1-207 aa encoded by BC006245)
FGF18 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. It has been shown in vitro that this protein is able to induce neurite outgrowth in PC12 cells. Studies of the similar proteins in mouse and chick suggested that this protein is a pleiotropic growth factor that stimulates proliferation in a number of tissues, most notably the liver and small intestine. Knockout studies of the similar gene in mice implied the role of this protein in regulating proliferation and differentiation of midline cerebellar structures.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGF; FGF-18; Fibroblast growth factor; Fibroblast growth factor 18; zFGF5
Gene Aliases: D130055P09Rik; FGF-18; FGF18; UNQ420/PRO856; ZFGF5
UniProt ID: (Human) O76093, (Mouse) O89101
Entrez Gene ID: (Human) 8817, (Mouse) 14172
Molecular Function: growth factor intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support