Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive test controls include: Mouse brain, Rat brain. The target is usually found in the following locations: Secreted.
Immunogen sequence: YPNASPLLGS SWGGLIHLYT ATARNSYHLQ IHKNGHVDGA PHQTIYSALM IRSEDAGFVV ITGVMSRRYL CMDFRGNIFG SHYFDPENCR FQHQTLENGY DVYHSPQYHF LVSLGRAKRA FLPGMNPPPY SQFLSRRNEI PLIHFNTPIP RRHTRSAEDD SERDPLNVLK PRARMTPAPA SCSQELPSAE DNSPMASDPL GVVRGGRVNT HAGGTGPEGC RPFAKFI
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The product of this gene inhibits renal tubular phosphate transport. This gene was identified by its mutations associated with autosomal dominant hypophosphatemic rickets, an inherited phosphate wasting disorder. Abnormally high level expression of this gene was found in oncogenic hypophosphatemic osteomalacia, a phenotypically similar disease caused by abnormal phosphate metabolism. Mutations in this gene have also been shown to cause familial tumoral calcinosis with hyperphosphatemia.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGF; FGF-23; Fibroblast growth factor; Fibroblast growth factor 23; Phosphatonin; tumor-derived hypophosphatemia inducing factor; Tumor-derived hypophosphatemia-inducing factor
Gene Aliases: ADHR; FGF23; FGFN; HPDR2; HYPF; PHPTC; UNQ3027/PRO9828
UniProt ID: (Human) Q9GZV9, (Mouse) Q9EPC2, (Rat) Q8VI82
Entrez Gene ID: (Human) 8074, (Mouse) 64654, (Rat) 170583
Molecular Function: growth factor intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support