Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is NIH-3T3 cell lysate.
PA5-20483 can be used with blocking peptide PEP-0603.
Fibroblast growth factor 4 (FGF4) is a member of the fibroblast growth factor (FGF) family that possess broad mitogenic and cell survival activities and play key roles in growth and survival of stem cells during embryogenesis, tissue regeneration, and carcinogenesis. FGF4 was identified by its strong oncogenic transforming activity and is a potent angiogenic factor, expressed in several highly vascularized tumors and also in adult mouse testis, intestine, and brain. Studies on the mouse homolog suggests a function in bone morphogenesis and limb development through the sonic hedgehog (SHH) signaling pathway. Furthermore, FGF4 regulates neural progenitor cell proliferation and neuronal differentiation. Recent studies show a growth-promoting role for FGF4 in human embryonic stem cells and a putative feedback inhibition mechanism by a novel FGF4 splice isoform that may serve to promote differentiation at a later stages of development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGF; FGF-4; Fibroblast growth factor; Fibroblast growth factor 4; fibroblast growth factor 4 splice isoform; HBGF-4; heparin secretory transforming protein 1; Heparin secretory-transforming protein 1; Heparin-binding growth factor 4; HST; HSTF-1; human stomach cancer, transforming factor from FGF-related oncogene; K-fibroblast growth factor; kaposi sarcoma oncogene; oncogene HST; Transforming protein KS3
Gene Aliases: Fgf-4; FGF4; Fgfk; HBGF-4; HST; HST-1; Hst1; Hstf-1; HSTF1; K-FGF; KFGF; KS3
UniProt ID: (Human) P08620, (Mouse) P11403
Entrez Gene ID: (Human) 2249, (Mouse) 14175
Molecular Function:
growth factor
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support