Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MAATAAAVV AEEDTELRDL LVQTLENSGV LNRIKAELRA AVFLALEEQE KVENKTPLVN ESLRKFLNTK DGRLVASLVA EFLQFFNLDF TLAVFQPETS TLQGLEGREN LARDLGIIEA EGTVGGPLLL EVIRRCQQKE KGPTTGEGAL DLSDVHSPPK SPEGKTSAQT TPSKKANDEA NQSDTSVSLS EPKSKSSLHL LSHETKIGSF LSNRTLDGKD KAGLCPDEDD MEGDSFFDDP IPKPEKTYGL RNEPRKQAGS LASLSDAPPL KSGLSSLAGA PSLKDSESKR GNTVLKDLKL ISDKIGSLGL GTGEDDDYVD DFNSTSHRSE KSEISIGEEI EEDLSVEIDD INTSDKLDDL TQDLTVSQLS DVADY (1-374 aa encoded by BC011902)
This gene encodes a largely hydrophilic protein postulated to be a leucine-rich protein family member. A t(6;8)(q27;p11) chromosomal translocation, fusing this gene and the fibroblast growth factor receptor 1 (FGFR1) gene, has been found in cases of myeloproliferative disorder. The resulting chimeric protein contains the N-terminal leucine-rich region of this encoded protein fused to the catalytic domain of FGFR1. This gene is thought to play an important role in normal proliferation and differentiation of the erythroid lineage. Alternatively spliced transcript variants that encode different proteins have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Centrosomal protein 43; FGFR1 oncogene partner; fibroblast growth factor receptor 1 oncogene partner
Gene Aliases: 4930553O10Rik; CEP43; FGFR1OP; FOP
UniProt ID: (Human) O95684, (Mouse) Q66JX5, (Rat) Q4V7C1
Entrez Gene ID: (Human) 11116, (Mouse) 75296, (Rat) 683722
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support