Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
FGFR4 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy. FGFR4 is required for normal down-regulation of the expression of CYP7A1, the rate-limiting enzyme in bile acid synthesis, in response to FGF19. FGFR4 plays a role in postnatal lung development and may be involved in the development of skeletal muscle cell lineages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD334; FGF Receptor; FGF Receptor 4; FGFR; FGFR-4; Fibroblast growth factor receptor 4; hydroxyaryl-protein kinase; MGC20292; protein-tyrosine kinase; tyrosine kinase related to fibroblast growth factor receptor; tyrosylprotein kinase
Gene Aliases: CD334; FGFR4; JTK2; TKF
UniProt ID: (Human) P22455
Entrez Gene ID: (Human) 2264
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support