Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: HepG2. The target is usually found in the following locations: Secreted.
Immunogen sequence: MAKVFSFILV TTALTMGREI SALEDCAQEQ MRLRAQVRLL ETRVKQQQVK IKQLLQENEV QFLDKGDENT VIDLGSKRQY ADCSEIFNDG YKLSGFYKIK PLQSPAEFSV YCDMSDGGGW TVIQRRSDGS ENFNRGWKDY ENGFGNFVQK HGEYWLGNKN LHFLTTQEDY TLKIDLADFE KNSRYAQYKN FKVGDEKNFY ELNIGEYSGT AGDSLAGNFH PEVQWWASHQ RMKFSTWDRD HDNYEGNCAE EDQSGWWFNR CHSANLNGVY YSGPYTAKTD NGIVWYTWHG WWYSLKSVVM KIRPNDFIPN VI
Fibrinogen-like 1 is a member of the fibrinogen family. This protein is homologous to the carboxy terminus of the fibrinogen beta- and gamma- subunits which contains the four conserved cysteines of fibrinogens and fibrinogen related proteins. However, this protein lacks the platelet-binding site, cross-linking region and a thrombin-sensitive site which are necessary for fibrin clot formation. This protein may play a role in the development of hepatocellular carcinomas. Four alternatively spliced transcript variants encoding the same protein exist for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fibrinogen-like 1; Fibrinogen-like protein 1; Hepassocin; hepatocellular carcinoma-related sequence; Hepatocyte-derived fibrinogen-related protein 1; HFREP-1; HP-041; HPS; LFIRE-1; Liver fibrinogen-related protein 1; MGC12455; OTTHUMP00000122468
Gene Aliases: FGL1; HFREP1; HP-041; LFIRE-1; LFIRE1
UniProt ID: (Human) Q08830
Entrez Gene ID: (Human) 2267
Molecular Function: intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support