Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This is a rabbit polyclonal antibody raised against full length of human FHL1.
Immunogen sequence: MAEKFDCHY CRDPLQGKKY VQKDGHHCCL KCFDKFCANT CVECRKPIGA DSKEVHYKNR FWHDTCFRCA KCLHPLANET FVAKDNKILC NKCTTREDSP KCKGCFKAIV AGDQNVEYKG TVWHKDCFTC SNCKQVIGTG SFFPKGEDFY CVTCHETKFA KHCVKCNKAI TSGGITYQDQ PWHADCFVCV TCSKKLAGQR FTAVEDQYYC VDCYKNFVAK KCAGCKNPIT GFGKGSSVVA YEGQSWHDYC FHCKKCSVNL ANKRFVFHQE QVYCPDCAKK L (1-280 aa encoded by BC010998)
This gene encodes a member of the four-and-a-half-LIM-only protein family. Family members contain two highly conserved, tandemly arranged, zinc finger domains with four highly conserved cysteines binding a zinc atom in each zinc finger. Expression of these family members occurs in a cell- and tissue-specific mode and these proteins are involved in many cellular processes. Mutations in this gene have been found in patients with Emery-Dreifuss muscular dystrophy. Multiple alternately spliced transcript variants which encode different protein isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA535K18.1; FHL; FHL-1; FHL1; FHL1B; formerly SLIM1; four and a half LIM domains a; Four and a half LIM domains protein 1; four-and-a-half Lin11, Isl-1 and Mec-3 domains 1; heart protein with four and a half LIM domains; KYO-T; KyoT; LIM protein SLIMMER; MGC111107; RAM14-1; RBP-associated molecule 14-1; RP11-535K18.1; skeletal and cardiac muscle SLIM isoform; Skeletal muscle LIM-protein 1; SLIM; unnamed protein product
Gene Aliases: cb1039; FCMSU; fhl; FHL-1; FHL1; FHL1A; FHL1B; fhla; FLH1A; KYOT; RAM14-1; RBMX1A; RBMX1B; SLIM; SLIM-1; SLIM1; SLIMMER; XMPMA; zgc:92025
UniProt ID: (Human) Q13642, (Rat) Q9WUH4, (Mouse) P97447
Entrez Gene ID: (Human) 2273, (Rat) 25177, (Mouse) 14199, (Zebrafish) 399646, (Zebrafish) 387528
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support