Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGVEIETIS PGDGRTFPKK GQTCVVHYTG MLQNGKKFDS SRDRNKPFKF RIGKQEVIKG FEEGAAQLGP LSPLPICPHP C (1-80 aa encoded by BC002614)
Has the potential to contribute to the immunosuppressive and toxic effects of FK506 and rapamycin. PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 12.6 kDa FK506-binding protein; 12.6 kDa FKBP; 126 kDa FKBP; calstabin 2; Calstabin-2; FK506 binding protein 1B, 12.6 kDa; FK506-binding protein 12.6; FK506-binding protein 1B; FKBP-12.6; FKBP-126; FKBP-1B; FKBP126; h-FKBP-12; Immunophilin FKBP12.6; Peptidyl-prolyl cis-trans isomerase FKBP1B; PPIase FKBP1B; Rotamase
Gene Aliases: 12.6kDa; AW494148; calstabin2; FKBP12.6; FKBP1B; FKBP1L; FKBP9; OTK4; PKBP1L; PPIase
UniProt ID: (Human) P68106, (Mouse) Q9Z2I2, (Rat) P97534
Entrez Gene ID: (Human) 2281, (Mouse) 14226, (Rat) 58950
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support