Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MAAAVPQRA WTVEQLRSEQ LPKKDIIKFL QEHGSDSFLA EHKLLGNIKN VAKTANKDHL VTAYNHLFET KRFKGTESIS KVSEQVKNVK LNEDKPKETK SEETLDEGPP KYTKSVLKKG DKTNFPKKGD VVHCWYTGTL QDGTVFDTNI QTSAKKKKNA KPLSFKVGVG KVIRGWDEAL LTMSKGEKAR LEIEPEWAYG KKGQPDAKIP PNAKLTFEVE LVDID (1-224 aa encoded by BC020809)
The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin, as well as histone deacetylases, the transcription factor YY1, casein kinase II, and nucleolin. It has a higher affinity for rapamycin than for FK506 and thus may be an important target molecule for immunosuppression by rapamycin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 25 kDa FK506-binding protein; 25 kDa FKBP; FK506 binding protein 3, 25kDa; FK506-binding protein 25, T-cell; FK506-binding protein 3; FK506-binding protein 3 (25kD); FKBP-25; FKBP-3; Immunophilin FKBP25; Peptidyl-prolyl cis-trans isomerase FKBP3; PPIase FKBP3; rapamycin binding protein; Rapamycin-selective 25 kDa immunophilin; Rotamase
Gene Aliases: 25kDa; FKBP-25; FKBP-3; FKBP25; FKBP3; PPIase
UniProt ID: (Human) Q00688, (Mouse) Q62446
Entrez Gene ID: (Human) 2287, (Mouse) 30795, (Rat) 299104
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support