Product References
Novel Oncogenic Transcription Factor Cooperation in RB-Deficient Cancer.
Cancer research
Mandigo AC,Shafi AA,McCann JJ,Yuan W,Laufer TS,Bogdan D,Gallagher L,Dylgjeri E,Semenova G,Vasilevskaya IA,Schiewer MJ,McNair CM,de Bono JS,Knudsen KE
PA1-020 was used in Western Blot to reveal that E2F1 binding is repositioned after retinoblastoma tumor suppressor loss to cooperate with androgen receptor and drive resultant gene expression networks that induce protumorigenic phenotypes.
Sat Jan 15 00:00:00 UTC 2022
Neuron-Derived Estrogen Is Critical for Astrocyte Activation and Neuroprotection of the Ischemic Brain.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Lu Y,Sareddy GR,Wang J,Zhang Q,Tang FL,Pratap UP,Tekmal RR,Vadlamudi RK,Brann DW
Published figure using FKBP5 polyclonal antibody (Product # PA1-020) in Western Blot
Wed Sep 16 00:00:00 UTC 2020
Neuron-Derived Estrogen Is Critical for Astrocyte Activation and Neuroprotection of the Ischemic Brain.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Lu Y,Sareddy GR,Wang J,Zhang Q,Tang FL,Pratap UP,Tekmal RR,Vadlamudi RK,Brann DW
Published figure using FKBP5 polyclonal antibody (Product # PA1-020) in Western Blot
Wed Sep 16 00:00:00 UTC 2020
Nucleocytoplasmic shuttling of the glucocorticoid receptor is influenced by tetratricopeptide repeat-containing proteins.
Journal of cell science
Mazaira GI,Echeverria PC,Galigniana MD
PA1-020 was used in Western Blotting to show that nuclear import of GR was impaired, whereas GR nuclear export was enhanced.
Tue Jun 16 00:00:00 UTC 2020
Heme Oxygenase 1 Impairs Glucocorticoid Receptor Activity in Prostate Cancer.
International journal of molecular sciences
Leonardi DB,Anselmino N,Brandani JN,Jaworski FM,Páez AV,Mazaira G,Meiss RP,Nuñez M,Nemirovsky SI,Giudice J,Galigniana M,Pecci A,Gueron G,Vazquez E,Cotignola J
PA1-020 was used in Western Blotting to identify the molecular processes governed by the interaction between haem oxygenase 1 and glucocorticoid receptor.
Tue Feb 26 00:00:00 UTC 2019
In silico identification and biochemical characterization of the human dicarboxylate clamp TPR protein interaction network.
FEBS open bio
Bernadotte A,Kumar R,Winblad B,Pavlov PF
PA1-020 was used in Western Blot to identify and characterise the human dicarboxylate clamp tetratricopeptide repeat protein interaction network.
Thu Nov 01 00:00:00 UTC 2018
Western High-Fat Diet Consumption during Adolescence Increases Susceptibility to Traumatic Stress while Selectively Disrupting Hippocampal and Ventricular Volumes.
eNeuro
Kalyan-Masih P,Vega-Torres JD,Miles C,Haddad E,Rainsbury S,Baghchechi M,Obenaus A,Figueroa JD
PA1020 was used in western blot to test if a Western-like high-fat diet predisposes rats to post-traumatic stress responsivity
Thu Oct 26 00:00:00 UTC 2017
Pharmacological stimulation of hypoxia inducible factor-1α facilitates the corticosterone response to a mild acute stressor.
Neuroscience letters
Harrell CS,Rowson SA,Neigh GN
PA1-020 was used in western blot to determine if HIF-1 links glucocorticoids and oxidative stress
Thu Jul 23 00:00:00 UTC 2015
Subcellular rearrangement of hsp90-binding immunophilins accompanies neuronal differentiation and neurite outgrowth.
Journal of neurochemistry
Quintá HR,Maschi D,Gomez-Sanchez C,Piwien-Pilipuk G,Galigniana MD
PA1-020 was used in immunocytochemistry and western blot to investigate the alterations hsp90-binding immunophilins distribution during neuronal growth and differentiation
Mon Nov 01 00:00:00 UTC 2010
Subcellular rearrangement of hsp90-binding immunophilins accompanies neuronal differentiation and neurite outgrowth.
Journal of neurochemistry
Quintá HR,Maschi D,Gomez-Sanchez C,Piwien-Pilipuk G,Galigniana MD
PA1-020 was used in immunocytochemistry and western blot to investigate the alterations hsp90-binding immunophilins distribution during neuronal growth and differentiation
Mon Nov 01 00:00:00 UTC 2010
PHLPP1 splice variants differentially regulate AKT and PKCα signaling in hippocampal neurons: characterization of PHLPP proteins in the adult hippocampus.
Journal of neurochemistry
Jackson TC,Verrier JD,Semple-Rowland S,Kumar A,Foster TC
PA1-020 was used in western blot to investigate the functions of different PHLPP splice variants in adult hippocampus
Mon Nov 01 00:00:00 UTC 2010
The hsp90-FKBP52 complex links the mineralocorticoid receptor to motor proteins and persists bound to the receptor in early nuclear events.
Molecular and cellular biology
Galigniana MD,Erlejman AG,Monte M,Gomez-Sanchez C,Piwien-Pilipuk G
PA1-020 was used in western blot to investigate the role of the heat shock protein 90-FKBP52 complex in the nucleus
Mon Mar 01 00:00:00 UTC 2010
Comparative analysis of calcineurin inhibition by complexes of immunosuppressive drugs with human FK506 binding proteins.
Biochemistry
Weiwad M,Edlich F,Kilka S,Erdmann F,Jarczowski F,Dorn M,Moutty MC,Fischer G
PA1-020 was used in western blot to study the inhibition of calcineurin by immunosuppressive drug-FK506 binding protein complexes
Tue Dec 26 00:00:00 UTC 2006
Ethanol-responsive brain region expression networks: implications for behavioral responses to acute ethanol in DBA/2J versus C57BL/6J mice.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Kerns RT,Ravindranathan A,Hassan S,Cage MP,York T,Sikela JM,Williams RW,Miles MF
PA1-020 was used in western blot to investigate the gene expression profile in brain regions of the mesolimbic reward pathway of mouse strains DBA/2J and C57BL/6J .
Wed Mar 02 00:00:00 UTC 2005
Up-regulation of glucocorticoid-regulated genes in a mouse model of Rett syndrome.
Human molecular genetics
Nuber UA,Kriaucionis S,Roloff TC,Guy J,Selfridge J,Steinhoff C,Schulz R,Lipkowitz B,Ropers HH,Holmes MC,Bird A
PA1-020 was used in western blot to study the expression of glucocorticoid-regulated genes in Rett syndrome mouse model.
Mon Aug 01 00:00:00 UTC 2005
Differential control of glucocorticoid receptor hormone-binding function by tetratricopeptide repeat (TPR) proteins and the immunosuppressive ligand FK506.
Biochemistry
Davies TH,Ning YM,Sánchez ER
PA1-020 was used in western blot to investigate the regulatory effects of FK506 and TPR proteins on glucocorticoid receptor hormone-binding function
Tue Feb 15 00:00:00 UTC 2005
Progesterone receptor deficient in chromatin binding has an altered cellular state.
The Journal of biological chemistry
Botos J,Xian W,Smith DF,Smith CL
PA1-020 was used in western blot to analyze the altered cellular processing by comparing the composition and distribution of tPR and sPR complexes
Fri Apr 09 00:00:00 UTC 2004
Association of immunophilins with mammalian TRPC channels.
The Journal of biological chemistry
Sinkins WG,Goel M,Estacion M,Schilling WP
PA1-020 was used in western blot to study the role of immunophilins in TRPC channel activation following receptor stimulation.
Fri Aug 13 00:00:00 UTC 2004
A new first step in activation of steroid receptors: hormone-induced switching of FKBP51 and FKBP52 immunophilins.
The Journal of biological chemistry
Davies TH,Ning YM,Sánchez ER
PA1-020 was used in western blot to investigate the activation mechanism for glucocorticoid receptors.
Fri Feb 15 00:00:00 UTC 2002
Tissue distribution and abundance of human FKBP51, and FK506-binding protein that can mediate calcineurin inhibition.
Biochemical and biophysical research communications
Baughman G,Wiederrecht GJ,Chang F,Martin MM,Bourgeois S
PA1-020 was used in western blot to study the expression of FK506-binding protein FKBP51 in tissues
Mon Mar 17 00:00:00 UTC 1997