Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • FKBP10 Antibodies

            Proteintech

            FKBP65 Polyclonal Antibody

            23 References
            View all (17) FKBP10 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite FKBP65 Polyclonal Antibody

            • Antibody Testing Data (12)
            Application
            FKBP65 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 12

            FKBP65 Antibody (12172-1-AP) in ICC/IF

            Immunofluorescent analysis of ( 10% Formaldehyde ) fixed HEK-293 cells using 12172-1-AP (FKBP65 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L).
            View Product

            FKBP65 Antibody in Immunocytochemistry (ICC/IF)
            FKBP65 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            FKBP65 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            FKBP65 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            FKBP65 Antibody in Western Blot (WB)
            FKBP65 Antibody in Western Blot (WB)
            FKBP65 Antibody in Western Blot (WB)
            FKBP65 Antibody in Western Blot (WB)
            FKBP65 Antibody in Western Blot (WB)
            FKBP65 Antibody in Western Blot (WB)
            FKBP65 Antibody in Western Blot (WB)
            FKBP65 Antibody in Immunoprecipitation (IP)

            Product Details

            12172-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            View 15 publications 15 publications

            Immunohistochemistry (IHC)

            -
            View 2 publications 2 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:20-1:200
            -

            Immunocytochemistry (ICC/IF)

            1:50-1:500
            View 6 publications 6 publications

            Immunoprecipitation (IP)

            0.5-4.0 µg
            -
            Product Specifications

            Species Reactivity

            Human, Mouse

            Published species

            Chicken, Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            FKBP65 Fusion Protein Ag2814 (233-582 aa encoded by BC016467)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.37 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: PFLAYGEKG YGTVIPPQAS LVFHVLLIDV HNPKDAVQLE TLELPPGCVR RAGAGDFMRY HYNGSLMDGT LFDSSYSRNH TYNTYIGQGY IIPGMDQGLQ GACMGERRRI TIPPHLAYGE NGTGDKIPGS AVLIFNVHVI DFHNPADVVE IRTLSRPSET CNETTKLGDF VRYHYNCSLL DGTQLFTSHD YGAPQEATLG ANKVIEGLDT GLQGMCVGER RQLIVPPHLA HGESGARGVP GSAVLLFEVE LVSREDGLPT GYLFVWHKDP PANLFEDMDL NKDGEVPPEE FSTFIKAQVS EGKGRLMPGQ DPEKTIGDMF QNQDRNQDGK ITVDELKLKS DEDEERVHEE L (233-582 aa encoded by BC016467)

            Target Information

            MAF acts as a transcriptional activator or repressor. The protein is involved in embryonic lens fiber cell development.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: 65 kDa FK506-binding protein; 65 kDa FKBP; FK506 binding protein 10, 65 kDa; FK506 binding protein 6 (65 kDa); FK506-binding protein 10; FKBP-10; FKBP-65; Immunophilin FKBP65; Peptidyl-prolyl cis-trans isomerase FKBP10; PPIase FKBP10; Rotamase

            View more View less

            Gene Aliases: AI325255; BRKS1; FKBP-10; FKBP-65; Fkbp-rs; Fkbp-rs1; Fkbp1-rs; FKBP10; Fkbp6; FKBP65; Fkbprp; hFKBP65; OI11; OI6; PPIASE; PSEC0056

            View more View less

            UniProt ID: (Human) Q96AY3, (Mouse) Q61576

            View more View less

            Entrez Gene ID: (Human) 60681, (Mouse) 14230

            View more View less

            Function(s)

            peptidyl-prolyl cis-trans isomerase activity calcium ion binding FK506 binding protein binding isomerase activity metal ion binding

            Process(es)

            protein peptidyl-prolyl isomerization chaperone-mediated protein folding protein folding
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0