Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FLAD1 encodes the enzyme that catalyzes adenylation of flavin mononucleotide (FMN) to form flavin adenine dinucleotide (FAD) coenzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FAD pyrophosphorylase; FAD synthase; FAD synthetase; FAD-synthetase; Fad1, flavin adenine dinucleotide synthetase, homolog; Flavin adenine dinucleotide synthase; flavin adenine dinucleotide synthetase; FMN adenylyltransferase; RP11-307C12.7
Gene Aliases: A930017E24Rik; FAD1; FADS; FLAD1; LSMFLAD; PP591
UniProt ID: (Human) Q8NFF5, (Mouse) Q8R123
Entrez Gene ID: (Human) 80308, (Mouse) 319945, (Rat) 751787
Molecular Function: hydrolase transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support