Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MWLDHRAVSQ VNRINETKHS VLQYVGGVMS VEMQAPKLLW LKENLREICW DKAGHFFDLP DFLSWKATGV TARSLCSLVC KWTYSAEKGW DDSFWKMIGL EDFVADNYSK IGNQVLPPGA SLGNGLTPEA ARDLGLLPGI AVAASLIDAH AGGLGVIGAD VRGHGLICEG QPVTSRLAVI CGTSSCHMGI SKDPIFVPGV WGPYFSAMVP GFWLNEGGQS VTGKLIDHMV QGHAAFPELQ VKATARCQSI YAYLNSHLDL IKKAQPVGFL TVDLHVWPDF HGNRSPLADL TLKGMVTGLK LSQDLDDLAI LYLATVQAIA LGTRFIIEAM EAAGHSISTL FLCGGLSKNP LFVQMHADIT GMPVVLSQEV ESVLVGAAVL GACASGDFAS VQEAMAKMSK VGKVVFPRLQ DKKYYDKKYQ VFLKLVEHQK EYLAIMNDD
FGGY gene ontology annotations related to this gene include neuron cellular homeostasis; pentose metabolic process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-ribulokinase FGGY; FGGY carbohydrate kinase domain-containing protein
Gene Aliases: FGGY
UniProt ID: (Human) Q96C11
Entrez Gene ID: (Human) 55277
Molecular Function:
carbohydrate kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support