Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TMA16 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Translation machinery-associated protein 16; UPF0534 protein C4orf43; UPF0534 protein C4orf43 homolog
Gene Aliases: 1810029B16Rik; C4orf43; RGD1305222; TMA16
UniProt ID: (Human) Q96EY4, (Mouse) Q9CR02
Entrez Gene ID: (Human) 55319, (Rat) 290686, (Mouse) 66282
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support