Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEKQPQNSRR GLAPREVPPA VGLLLIMALM NTLLYLCLDH FFIAPRQSTV DPTHCPYGHF RIGQMKNCSP WLSCEELRTE VRQLKRVGEG AVKRVFLSEW KEHKVALSQL TSLEMKDDFL HGLQMLKSLQ GTHVVTLLGY CEDDNTMLTE YHPLGSLSNL EETLNLSKYQ NVNTWQHRLE LAMDYVSIIN YLHHSPVGTR VMCDSNDLPK TLSQYLLTSN FSILANDLDA LPLVNHSSGM LVKCGHRELH GDFVAPEQLW PYGEDVPFHD DLMPSYDEKI DIWKIPDISS FLLGHIEGSD MVRFHLFDIH KACKSQTPSE RPTAQDVLET YQKVLDTLRD AMMSQAREML
This gene encodes a protein that may be involved in the presentation of the laminin-binding O-linked carbohydrate chain of alpha-dystroglycan (a-DG), which forms transmembrane linkages between the extracellular matrix and the exoskeleton. Some pathogens use this O-linked carbohydrate unit for host entry. Loss of function compound heterozygous mutations in this gene were found in a human patient affected by the Walker-Warburg syndrome (WWS) phenotype. Mice lacking this gene contain misplaced neurons (heterotopia) in some regions of the brain, possibly from defects in neuronal migration. Alternative splicing of this gene results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: POMK; probable inactive protein kinase-like protein SgK196; Protein kinase-like protein SgK196; Protein O-mannose kinase; Sugen kinase 196
Gene Aliases: MDDGA12; MDDGC12; POMK; SGK196
UniProt ID: (Human) Q9H5K3
Entrez Gene ID: (Human) 84197
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support