Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AQEEAYREEQ LINRLMRQSQ QERRIAVQLM HVRHEKEVLW QNRIFREKQH EERRLKDFQD ALDREAALAK QAKIDFEEQF LKEKRFHDQI AVERAQARY
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 122; Protein KPL2; Sperm flagellar protein 2; testis tissue sperm-binding protein Li 47a
Gene Aliases: CT122; KIAA1770; KPL2; SPEF2
UniProt ID: (Human) Q9C093
Entrez Gene ID: (Human) 79925
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support