Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGAVSCRQGQ HTQQGEHTRV AVPHKQGGNI RGPWARGWKS LWTGLGTIRS DLEELWELRG HHYLHQESLK PAPVLVEKPL PEWPVPQFIN LFLPEFPIRP IRGQQQLKIL GLVAKGSFGT VLKVLDCTQK AVFAVKVVPK VKVLQRDTVR QCKEEVSIQR QINHPFVHSL GDSWQGKRHL FIMCSYCSTD LYSLWSAVGC FPEASIRLFA AELVLVLCYL HDLGIMHRDV KMENILLDER GHLKLTDFGL SRHVPQGAQA YTICGTLQYM GERG
FLJ25006 likely exerts a protein serine/threonine kinase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: sugen kinase 494
Entrez Gene ID: (Human) 124923
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support