Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALEAAGGPP EETLSLWKRE QARLKAHVVD RDTEAWQRDP AFSGLQRVGG VDVSFVKGDS VRACASLVVL SFPELEVLLV DGNGVLHHRG FGVACHLGVL TDLPCVGVAK KLLQVDGLEN NALHKEKIRL LQTRGDSFPL LGDSGTVLGM ALRSHDRSTR PLYISVGHRM SLEAAVRLTC CCCRFRIPEP VRQADICSRE HIRKSLGLPG PPTPRSPKAQ RPVACPKGDS GESSGEGQPP QDHSPGPRTA PRPGSQEQAG KDWQ
ENDOV gene ontology annotations related to this gene include DNA repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endonuclease V; hEndoV; Inosine-specific endoribonuclease
Gene Aliases: ENDOV
UniProt ID: (Human) Q8N8Q3
Entrez Gene ID: (Human) 284131
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support