Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MARPDDEEGA AVAPGHPLAK GYLPLPRGAP VGKESVELQN GPKAGTFPVN GAPRDSLAAA SGVLGGPQTP LAPEEETQAR LLP
This gene encodes a member of the major facilitator superfamily of transporter proteins. The encoded protein is a heme transporter that may play a critical role in erythropoiesis by protecting developing erythroid cells from heme toxicity. This gene may play a role in posterior column ataxia with retinitis pigmentosa and the hematological disorder Diamond-Blackfan syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Choline/ethanolamine transporter FLVCR1; Feline leukemia virus subgroup C receptor; Feline leukemia virus subgroup C receptor-related protein 1; FLVCR protein; Heme transporter FLVCR1
Gene Aliases: AXPC1; FLVCR; FLVCR1; MFSD7B; PCA; PCARP
UniProt ID: (Human) Q9Y5Y0
Entrez Gene ID: (Human) 28982
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support