Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Plays a role in the formation of adherens junction and the polymerization of linear actin cables.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: formin (limb deformity); formin 1; Formin-1; Limb deformity protein; Limb deformity protein homolog
Gene Aliases: FMN; FMN1; LD; RGD1306349; RGD1310100
UniProt ID: (Human) Q3B7I6, (Mouse) Q05860
Entrez Gene ID: (Human) 342184, (Rat) 296512, (Mouse) 14260
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support