Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Peptide sequence: EKQLAQEAKK LDAKTPSQRN KWQQQELIAE LRRRQAKEHR PVYEGKDGTI
FMNL3, also known as Formin-like protein 3, is a 1028 amino acid containing protein involved in immunity or defense. This developmental protein is implicated in polarity control, invasion, migration, or metastasis through regulation of the Rho-related signaling pathway. It belongs to the formin homology family and contains a FH2 (formin homology 2) domain and a GBD/FH3 (Rho GTPase-binding/formin homology 3) domain and has high sequence identity to the mouse Wbp3 protein. Two different isoforms have been described. Itis suspectedbe involved in gastric cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Formin homology 2 domain-containing protein 3; formin-like 3; Formin-like protein 3; WBP-3; WW domain binding protein 3; WW domain-binding protein 3
Gene Aliases: FHOD3; FMNL3; FRL2; KIAA2014; WBP-3; WBP3
UniProt ID: (Human) Q8IVF7
Entrez Gene ID: (Human) 91010
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support