Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLKHIQFKTK VCSVTKCSDS AVSGQWEVVT MHEEKQESAI FDAVMVCTGF LTNPYLPLDS FPGINAFKGQ YFHSRQYKHP DIFKDKRVLV IGMGNSGTDI AVEASHLAEK VFLSTTGGGW VISRIFDSGY PWDMVFMTRF QNMLRNSLPT PIVTWLMERK INNWLNHANY GLIPEDRTQL KEFVLNDELP GRIITGKVFI RPSIKEVKEN SVIFNNTSKE EPIDIIVFAT GYTFAFPFLD ESVVKVEDGQ ASLYKYIFPA HLQKPTLAII GLIKPLGSMI PTGETQARWA VRVLKGVNKL PPPSVMIEEI NARKENKPSW FGLCYCKALQ S
Metabolic N-oxidation of the diet-derived amino-trimethylamine (TMA) is mediated by flavin-containing monooxygenase and is subject to an inherited FMO3 polymorphism in man resulting in a small subpopulation with reduced TMA N-oxidation capacity resulting in fish odor syndrome Trimethylaminuria. Three forms of the enzyme, FMO1 found in fetal liver, FMO2 found in adult liver, and FMO3 are encoded by genes clustered in the 1q23-q25 region. Flavin-containing monooxygenases are NADPH-dependent flavoenzymes that catalyzes the oxidation of soft nucleophilic heteroatom centers in drugs, pesticides, and xenobiotics.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 1; Dimethylaniline oxidase 1; Fetal hepatic flavin-containing monooxygenase 1; Flavin-containing monooxygenase 1; Flavin-containing monooxygenase 1 (fetal liver); FMO 1; Hepatic flavin-containing monooxygenase 1; Trimethylamine monooxygenase
Gene Aliases: Fmo-1; FMO1
UniProt ID: (Human) Q01740, (Mouse) P50285
Entrez Gene ID: (Human) 2326, (Mouse) 14261
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support