Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to FMO3.
Immunogen sequence: EPTCFEKSN DIGGLWKFSD HAEEGRASIY KSVFSNSSKE MMCFPDFPFP DDFPNFMHNS KIQEYIIAFA KEKNLLKYIQ FKTFVSSVNK HPDFATTGQW DVTTERDGKK ESAVFDAVMV CSGHHVYPNL PKESFPGLNH FKGKCFHSRD YKEPGVFNGK RVLVVGLGNS GCDIATELSR TAEQVMISSR SGSWVMSRVW DNGYPWDMLL VTRFGTFLKN NLPTAISDWL YMKQMNARFK HENYGLMPLN GVLRKEPVFN DELPASILCG IVSVKPNVKE FTETSAIFED GTIFEGIDCV IFATGYSFAY PFLDESIIKS RNNEII (27-351 aa encoded by BC032016)
The mammalian flavin-containing monooxygenases (FMO; EC 1.14.13.8) represent a multigene family whose gene products are localized in the endoplasmic reticulum of many tissues. These enzymes catalyze the NADPH-dependent oxidative metabolism of many drugs, pesticides, and other foreign compounds. Their substrates are soft nucleophiles with an electron-rich center, typically a nitrogen, sulfur or phosphorus-containing functional group, as the site for oxidative attack by the enzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 3; Dimethylaniline oxidase 3; Flavin-containing monooxygenase 3; FMO 3; FMO form 2; FMO II; Hepatic flavin-containing monooxygenase 3; hepatic flavin-containing monooxygenase-3; MGC34400; Trimethylamine monooxygenase
Gene Aliases: AW111792; dJ127D3.1; FMO3; FMOII; TMAU
UniProt ID: (Human) P31513, (Rat) Q9EQ76, (Mouse) P97501
Entrez Gene ID: (Human) 2328, (Rat) 84493, (Mouse) 14262
Molecular Function: oxidoreductase oxygenase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support