Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The mammalian flavin-containing monooxygenases (FMO; EC 1.14.13.8) represent a multigene family whose gene products are localized in the endoplasmic reticulum of many tissues. These enzymes catalyze the NADPH-dependent oxidative metabolism of many drugs, pesticides, and other foreign compounds. Their substrates are soft nucleophiles with an electron-rich center, typically a nitrogen, sulfur or phosphorus-containing functional group, as the site for oxidative attack by the enzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 3; Dimethylaniline oxidase 3; Flavin-containing monooxygenase 3; FMO 3; FMO form 2; FMO II; Hepatic flavin-containing monooxygenase 3; hepatic flavin-containing monooxygenase-3; MGC34400; Trimethylamine monooxygenase
Gene Aliases: dJ127D3.1; FMO3; FMOII; TMAU
UniProt ID: (Human) P31513
Entrez Gene ID: (Human) 2328
Molecular Function:
oxidoreductase
oxygenase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support