Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody, generated by N-330 amino-acids of FMO4, can recognize the 2 variants and also cross-react with other FMO proteins.
Immunogen sequence: EDLEPTCFE RSDDIGGLWK FTESSKDGMT RVYKSLVTNV CKEMSCYSDF PFHEDYPNFM NHEKFWDYLQ EFAEHFDLLK YIQFKTTVCS ITKRPDFSET GQWDVVTETE GKQNRAVFDA VMVCTGHFLN PHLPLEAFPG IHKFKGQILH SQEYKIPEGF QGKRVLVIGL GNTGGDIAVE LSRTAAQVLL STRTGTWVLG RSSDWGYPYN MMVTRRCCSF IAQVLPSRFL NWIQERKLNK RFNHEDYGLS ITKGKKAKFI VNDELPNCIL CGAITMKTSV IEFTETSAVF EDGTVEENID VVIFTTGYTF SFPFFEEPLK SLCTKKIFLY KQ (24-354 aa encoded by BC002780)
FMO4 (Flavin-containing monooxygenase 4) also known as FMO2, FORMERLY or FMO, ADULT LIVER FORM, is an enzyme that in humans is encoded by the FMO4 gene. By PCR analysis, Dolphin et al. (1992) mapped the FMO4 gene to chromosome 1. Southern blot hybridization with single exon probes demonstrated that human FMO4 and FMO1 are, in each case, the product of a single gene. Lawton et al. (1994) proposed a reclassification of mammalian FMOs. Under this system, FMO2 described by Dolphin et al. (1992) is now designated FMO4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 4; Dimethylaniline oxidase 4; flavin-containing monooxygenase 4; FMO 4; Hepatic flavin-containing monooxygenase 4
Gene Aliases: D1Ertd532e; FMO2; FMO4
UniProt ID: (Human) P31512, (Mouse) Q8VHG0, (Rat) Q8K4B7
Entrez Gene ID: (Human) 2329, (Mouse) 226564, (Rat) 246247
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support