Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EDLEPTCFER SDDIGGLWKF TESSKDGMTR VYKSLVTNVC KEMSCYSDFP FHEDYPNFMN HEKFWDYLQE FAEHFDLLKY IQFKTTVCSI TKRPDFSETG QWDVVTETEG KQNRAVFDAV MVCTGHFLNP HLPLEAFPGI HKFKGQILHS QEYKIPEGFQ GKRVLVIGLG NTGGDIAVEL SRTAAQVLLS TRTGTWVLGR SSDWGYPYNM MVTRRCCSFI AQVLPSRFLN WIQERKLNKR FNHEDYGLSI TKGKKAKFIV NDELPNCILC GAITMKTSVI EFTETSAVFE DGTVEENIDV VIFTTGYTFS FPFFEEPLKS LCTKKIFLYK Q
FMO4 (Flavin-containing monooxygenase 4) also known as FMO2, FORMERLY or FMO, ADULT LIVER FORM, is an enzyme that in humans is encoded by the FMO4 gene. By PCR analysis, Dolphin et al. (1992) mapped the FMO4 gene to chromosome 1. Southern blot hybridization with single exon probes demonstrated that human FMO4 and FMO1 are, in each case, the product of a single gene. Lawton et al. (1994) proposed a reclassification of mammalian FMOs. Under this system, FMO2 described by Dolphin et al. (1992) is now designated FMO4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 4; Dimethylaniline oxidase 4; flavin containing monooxygenase 4; flavin-containing monooxygenase 4; flavin-containing monooxygenase FMO4; FMO 1E1; FMO 4; Hepatic flavin-containing monooxygenase 4
Gene Aliases: D1Ertd532e; FMO2; FMO4
UniProt ID: (Human) P31512, (Rabbit) P36367, (Mouse) Q8VHG0, (Rat) Q8K4B7
Entrez Gene ID: (Human) 2329, (Pig) 100621718, (Rabbit) 100009582, (Mouse) 226564, (Rat) 246247
Molecular Function: oxidoreductase oxygenase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support