Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RTDDIGGLW RFQENPEEGR ASIYKSVIIN TSKEMMCFSD YPIPDHYPNF MHNAQVLEYF RMYAKEFDLL KYIRFKTTVC SVKKQPDFAT SGQWEVVTES EGKKEMNVFD GVMVCTGHHT NAHLPLESFP GIEKFKGQYF HSRDYKNPEG FTGKRVIIIG IGNSGGDLAV EISQTAKQVF LSTRRGAWIL NRVGDYGYPA DVLFSSRLTH FIWKICGQSL ANKYLEKKIN QRFDHEMFGL KPKHRSKDIA LTE (34-285 aa encoded by BC035687)
Metabolic N-oxidation of the diet-derived amino-trimethylamine (TMA) is mediated by flavin-containing monooxygenase and is subject to an inherited FMO3 polymorphism in man resulting in a small subpopulation with reduced TMA N-oxidation capacity resulting in fish odor syndrome Trimethylaminuria. Three forms of the enzyme, FMO1 found in fetal liver, FMO2 found in adult liver, and FMO3 are encoded by genes clustered in the 1q23-q25 region. Flavin-containing monooxygenases are NADPH-dependent flavoenzymes that catalyzes the oxidation of soft nucleophilic heteroatom centers in drugs, pesticides, and xenobiotics. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Baeyer-Villiger monooxygenase 1; Dimethylaniline monooxygenase [N-oxide-forming] 5; Dimethylaniline oxidase 5; Flavin-containing monooxygenase 5; FMO 5; hBVMO1; Hepatic flavin-containing monooxygenase 5; NADPH oxidase
Gene Aliases: 5033418D19Rik; AI195026; FMO5
UniProt ID: (Human) P49326, (Rat) Q8K4C0, (Mouse) P97872
Entrez Gene ID: (Human) 2330, (Rat) 246248, (Mouse) 14263
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support