Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Centrosomal protein 20; FGFR1OP N-terminal-like protein; FOP-related protein of 20 kDa; lisH domain-containing protein C16orf63; lisH domain-containing protein C16orf63 homolog; LisH domain-containing protein FOPNL; pluripotent embryonic stem cell-related protein
Gene Aliases: 0610037P05Rik; C16orf63; CEP20; FOPNL; FOR20; PHSECRG2; RGD1305823
UniProt ID: (Human) Q96NB1, (Mouse) Q9CZS3
Entrez Gene ID: (Human) 123811, (Mouse) 66086, (Rat) 360461
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support