Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: QPMRSGGGSV GAVVVKQEPL EEDSPSSSSA GLDKAQRSVI KPISIAGGFY GEEPLHTPIV VTSTPAVTPG TSNLVFTYPS VLEQESPASP
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FOS like 2, AP-1 trancription factor subunit; FOS like antigen 2; FOS-like antigen 2; Fos-related antigen 2; FRA-2
Gene Aliases: FOSL2; FRA2
UniProt ID: (Human) P15408
Entrez Gene ID: (Human) 2355
Molecular Function: basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support