Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KCEKQPGAG GGGGSGSGGS GAKGGPESRK DPSGASNPSA DSPLHRGVHG KTGQLEGAPA PGPAASPQTL DHSGATATGG ASELKTPASS TAPPISSGPG ALASVPASHP AHGLAPHESQ LHLKGDPHYS FNHP (267-399 aa encoded by B C033890)
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ci-fkh; fkd7; fkh; Forkhead box protein A1; forkhead-7; FoxA4a; Hepatocyte nuclear factor 3-alpha; HNF-3-alpha; HNF-3A; HNF3-alpha; HNF3alpha; MGC33105; MGC52590; MGC75967; pintallavis; TCF-3A; tFoxA1; Transcription factor 3A; xfkh1; xfkh2
Gene Aliases: FOXA1; HNF3A; TCF3A
UniProt ID: (Human) P55317
Entrez Gene ID: (Human) 3169
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support