Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total FOXB1/2.
The Forkhead-box (FOX) genes comprise a superfamily of at least 43 members that encode proteins which are involved in transcriptional regulation and may be associated with the pathogenesis of various cancers. FOXB1 (forkhead box B1), also known as FKH5 or HFKH-5, and FOXB2 (forkhead box B2) are members of the FOX family and each contain one forkhead DNA-binding domain. Both FOXB1 and FOXB2 localize to the nucleus where they are thought to function as transcription factors that can bind to DNA via their forkhead domains. In mice, defects in the gene encoding FOXB1 are associated with retarded development of the central nervous system (CNS), suggesting that FOXB1 may play a role in CNS organization and function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: forkhead box B1a; forkhead box B1b; Forkhead box protein B1; Forkhead box protein B2; forkhead homolog 4; mesoderm/mesenchyme forkhead 3; Transcription factor FKH-4; Transcription factor FKH-5
Gene Aliases: bA159H20.4; C43; Fkh4; FKH5; FOXB1; Foxb1a; Foxb1b; FOXB2; Hfh-e5.1; HFKH-5; Mf3; Twh
UniProt ID: (Human) Q99853, (Human) Q5VYV0, (Mouse) Q64732, (Mouse) Q64733
Entrez Gene ID: (Human) 27023, (Human) 442425, (Mouse) 64290, (Mouse) 14240
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support