Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Foxb1 gene ontology annotations related to this gene include anatomical structure morphogenesis; axon target recognition; cell differentiation; cell migration in diencephalon; epithelial cell differentiation involved in mammary gland alveolus development; floor plate development; hypothalamus cell migration; inferior colliculus development; lactation; mammary gland lobule development; mammillary body development; mammillothalamic axonal tract development; midbrain development; negative regulation of neuron apoptotic process; regulation of transcription by RNA polymerase II; somitogenesis; spinal cord development; telencephalon cell migration; thalamus development; urogenital system development; visual learning.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Forkhead box protein B1; Transcription factor FKH-5
Gene Aliases: FKH5; FOXB1; HFKH-5
UniProt ID: (Human) Q99853
Entrez Gene ID: (Human) 27023
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support