Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Forkhead box protein C2 (FOXC2) is a transciptional activator and is characterized by a distinct DNA-binding forkhead domain. FOXC2 may be involved in the formation of special mesenchymal tissues. Mutations affecting the gene can cause lymphedema-distichiasis (LYD).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: forkhead box C2 (MFH-1, mesenchyme forkhead 1); Forkhead box protein C2; forkhead, Drosophila, homolog-like 14; Forkhead-related protein FKHL14; Mesenchyme fork head protein 1; mesenchyme forkhead 1; MFH-1 protein; MFH-1,mesenchyme forkhead 1; Transcription factor FKH-14
Gene Aliases: FKHL14; FOXC2; LD; MFH-1; MFH1
UniProt ID: (Human) Q99958
Entrez Gene ID: (Human) 2303
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support