Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
FOXH1 is a member of the Forkhead-box (FOX) family of transcription factors and is the human homolog of Xenopus forkhead activin signal transducer-1. FOXH1 plays an important role in mediating TGF-beta/activin signals through its interaction with the SMAD2-SMAD4 complex. It is expressed primarily during early development and localizes in the cytoplasm until it forms a complex with the SMAD proteins at which time it translocates to the nucleus to positively or negatively affect gene expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: contains winged-helix domain; etID62177.12; Fast-1; Forkhead activin signal transducer 1; Forkhead box protein H1; Schmalspur protein; transcription factor; uncle freddy; unf
Gene Aliases: fast; fast1; fe26b06; foxh1; schmalspur; SUR; wu:fe26b06; wu:fi30c08; wu:fi32b01
UniProt ID: (Zebrafish) Q9I9E1
Entrez Gene ID: (Zebrafish) 57930
Molecular Function: winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support