Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: HLFDPRLPAL AASPMLYLYG PERPGLPLAF APAAALAASG RAETPQKPPY
Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 92%; Rat: 86%
The forkhead domain is a 100-amino acid monomeric DNA binding motif originally identified as a region of homology between the Drosophila forkhead protein and rat HNF3. Pierrou et al. (1994) identified 7 human genes containing forkhead domains and designated them forkhead related activators (FREAC) 1 through 7.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FKH11; FKHL10; Forkhead box protein L1; forkhead homolog 6; Forkhead related transcription factor; forkhead-like 11; Forkhead-related protein FKHL11; Forkhead-related transcription factor 7; FREAC-7; FREAC6; JFJ-3; Transcription factor FKH-6
Gene Aliases: fkh-6; FKH6; FKHL11; FOXL1; FREAC7
UniProt ID: (Human) Q12952, (Mouse) Q64731
Entrez Gene ID: (Human) 2300, (Mouse) 14241
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support