Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Antibody is stable for 24 months.
Positive Control: HeLa or U87 cells.Ubiquitous tissue expression. Cellular Localization: Nucleus.
Specificity Comments: FOXL1 is a 337 amino acid protein encoded by the mouse gene FOXL1. FOXL1 belongs to the forkhead family and contains one forkhead DNA-binding domain. The HNF3/forkhead family includes a large number of transcription factors that share a structurally related DNA binding domain. forkhead factors are known to play important roles both during development and in adults. FOXL1 is a winged helix transcriptional regulator expressed in the mesenchymal layer of developing and mature gastrointestinal tract. FOXL1-deficient mice exhibit various defects not only in the epithelial layer of the gastrointestinal tract but also in gut-associated lymphoid tissues. In the small intestine of FOXL1-deficient mice, the formation of Peyers patches is affected, particularly in the caudal region. FOXL1 is a mesenchymal modifier of the Adenomatous Polyposis Coli (APC) gene products and plays a key role in gastrointestinal tumorigenesis.
Predicted to react in Rat
The forkhead domain is a 100-amino acid monomeric DNA binding motif originally identified as a region of homology between the Drosophila forkhead protein and rat HNF3. Pierrou et al. (1994) identified 7 human genes containing forkhead domains and designated them forkhead related activators (FREAC) 1 through 7.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FKH11; FKHL10; Forkhead box protein L1; Forkhead related transcription factor; forkhead-like 11; Forkhead-related protein FKHL11; Forkhead-related transcription factor 7; FREAC-7; FREAC6; JFJ-3
Gene Aliases: FKH6; FKHL11; FOXL1; FREAC7
UniProt ID: (Human) Q12952
Entrez Gene ID: (Human) 2300
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support