Product References
Vitamin D3 mitigates autoimmune inflammation caused by activation of myeloid dendritic cells in SLE.
Experimental dermatology
Li M,Luo L,Lin C,Ni B,Zou L,Song Z,Hao F,Wu Y,Luo N
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Mon Jan 01 00:00:00 UTC 2024
Changes of T Lymphocyte Subsets in Peripheral Blood of Patients with Intermediate and Advanced Cervical Cancer before and after Nimotuzumab Combined with Chemoradiotherapy.
International archives of allergy and immunology
Teng F,Cui G,Qian L,Zhao L
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Thu Jan 05 00:00:00 UTC 2023
Tumor microenvironment shows an immunological abscopal effect in patients with NSCLC treated with pembrolizumab-radiotherapy combination.
Journal for immunotherapy of cancer
van der Woude LL,Gorris MAJ,Wortel IMN,Creemers JHA,Verrijp K,Monkhorst K,Grünberg K,van den Heuvel MM,Textor J,Figdor CG,Piet B,Theelen WSME,de Vries IJM
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Sat Oct 01 00:00:00 UTC 2022
Characterizing CDK12-Mutated Prostate Cancers.
Clinical cancer research : an official journal of the American Association for Cancer Research
Rescigno P,Gurel B,Pereira R,Crespo M,Rekowski J,Rediti M,Barrero M,Mateo J,Bianchini D,Messina C,Fenor de la Maza MD,Chandran K,Carmichael J,Guo C,Paschalis A,Sharp A,Seed G,Figueiredo I,Lambros M,Miranda S,Ferreira A,Bertan C,Riisnaes R,Porta N,Yuan W,Carreira S,de Bono JS
13-4777 was used in Immunohistochemistry-immunofluorescence to characterize CDK12-mutated mCRPC, presenting clinical, genomic, and tumor-infiltrating lymphocyte (TIL) data.
Fri Jan 15 00:00:00 UTC 2021
Analysis of Expression of Different Histone Deacetylases in Autoimmune Thyroid Disease.
The Journal of clinical endocrinology and metabolism
Sacristán-Gómez P,Serrano-Somavilla A,González-Amaro R,Martínez-Hernández R,Marazuela M
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Thu Oct 21 00:00:00 UTC 2021
Common clonal origin of conventional T cells and induced regulatory T cells in breast cancer patients.
Nature communications
Xydia M,Rahbari R,Ruggiero E,Macaulay I,Tarabichi M,Lohmayer R,Wilkening S,Michels T,Brown D,Vanuytven S,Mastitskaya S,Laidlaw S,Grabe N,Pritsch M,Fronza R,Hexel K,Schmitt S,Müller-Steinhardt M,Halama N,Domschke C,Schmidt M,von Kalle C,Schütz F,Voet T,Beckhove P
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Thu Feb 18 00:00:00 UTC 2021
Increased CD3+, CD8+, or FoxP3+ T Lymphocyte Infiltrations Are Associated with the Pathogenesis of Colorectal Cancer but Not with the Overall Survival of Patients.
Biology
Barbosa AM,Martinho O,Nogueira R,Campos J,Lobo L,Pinto H,Longatto-Filho A,Castro AG,Martins SF,Torrado E
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Fri Aug 20 00:00:00 UTC 2021
Multiple Immunostainings with Different Epitope Retrievals-The FOLGAS Protocol.
International journal of molecular sciences
von Schoenfeld A,Bronsert P,Poc M,Fuller A,Filby A,Kraft S,Kurowski K,Sörensen K,Huber J,Pfeiffer J,Proietti M,Stehl V,Werner M,Seidl M
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Sat Dec 25 00:00:00 UTC 2021
Immune cell topography predicts response to PD-1 blockade in cutaneous T cell lymphoma.
Nature communications
Phillips D,Matusiak M,Gutierrez BR,Bhate SS,Barlow GL,Jiang S,Demeter J,Smythe KS,Pierce RH,Fling SP,Ramchurren N,Cheever MA,Goltsev Y,West RB,Khodadoust MS,Kim YH,Schürch CM,Nolan GP
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Thu Nov 18 00:00:00 UTC 2021
Muscle-Directed Delivery of an AAV1 Vector Leads to Capsid-Specific T Cell Exhaustion in Nonhuman Primates and Humans.
Molecular therapy : the journal of the American Society of Gene Therapy
Gernoux G,Gruntman AM,Blackwood M,Zieger M,Flotte TR,Mueller C
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Wed Mar 04 00:00:00 UTC 2020
Mitochondrial transfer from MSCs to T cells induces Treg differentiation and restricts inflammatory response.
EMBO reports
Court AC,Le-Gatt A,Luz-Crawford P,Parra E,Aliaga-Tobar V,Bátiz LF,Contreras RA,Ortúzar MI,Kurte M,Elizondo-Vega R,Maracaja-Coutinho V,Pino-Lagos K,Figueroa FE,Khoury M
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Wed Feb 05 00:00:00 UTC 2020
Single-cell multi-omics analysis presents the landscape of peripheral blood T-cell subsets in human chronic prostatitis/chronic pelvic pain syndrome.
Journal of cellular and molecular medicine
Zhang M,Liu Y,Chen J,Chen L,Meng J,Yang C,Yin S,Zhang X,Zhang L,Hao Z,Chen X,Liang C
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Tue Dec 01 00:00:00 UTC 2020
hESC-derived immune suppressive dendritic cells induce immune tolerance of parental hESC-derived allografts.
EBioMedicine
Todorova D,Zhang Y,Chen Q,Liu J,He J,Fu X,Xu Y
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Tue Dec 01 00:00:00 UTC 2020
Biomarker recommendation for PD-1/PD-L1 immunotherapy development in pediatric cancer based on digital image analysis of PD-L1 and immune cells.
The journal of pathology. Clinical research
Silva MA,Triltsch N,Leis S,Kanchev I,Tan TH,Van Peel B,Van Kerckhoven M,Deschoolmeester V,Zimmermann J
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Wed Apr 01 00:00:00 UTC 2020
Digital Pathology Analysis Quantifies Spatial Heterogeneity of CD3, CD4, CD8, CD20, and FoxP3 Immune Markers in Triple-Negative Breast Cancer.
Frontiers in physiology
Mi H,Gong C,Sulam J,Fertig EJ,Szalay AS,Jaffee EM,Stearns V,Emens LA,Cimino-Mathews AM,Popel AS
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Wed Nov 18 00:00:00 UTC 2020
Distinct Tumor Microenvironments Are a Defining Feature of Strain-Specific CRISPR/Cas9-Induced MPNSTs.
Genes
Scherer A,Stephens VR,McGivney GR,Gutierrez WR,Laverty EA,Knepper-Adrian V,Dodd RD
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Sat May 23 00:00:00 UTC 2020
Inhibition of RANK signaling in breast cancer induces an anti-tumor immune response orchestrated by CD8+ T cells.
Nature communications
Gómez-Aleza C,Nguyen B,Yoldi G,Ciscar M,Barranco A,Hernández-Jiménez E,Maetens M,Salgado R,Zafeiroglou M,Pellegrini P,Venet D,Garaud S,Trinidad EM,Benítez S,Vuylsteke P,Polastro L,Wildiers H,Simon P,Lindeman G,Larsimont D,Van den Eynden G,Velghe C,Rothé F,Willard-Gallo K,Michiels S,Muñoz P,Walzer T,Planelles L,Penninger J,Azim HA,Loi S,Piccart M,Sotiriou C,González-Suárez E
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Thu Dec 10 00:00:00 UTC 2020
Inhibitory activity of FOXP3+ regulatory T cells reveals high specificity for displaying immune tolerance in remission state rheumatoid arthritis.
Scientific reports
Kanjana K,Chevaisrakul P,Matangkasombut P,Paisooksantivatana K,Lumjiaktase P
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Fri Nov 13 00:00:00 UTC 2020
Hijacking antibody-induced CTLA-4 lysosomal degradation for safer and more effective cancer immunotherapy.
Cell research
Zhang Y,Du X,Liu M,Tang F,Zhang P,Ai C,Fields JK,Sundberg EJ,Latinovic OS,Devenport M,Zheng P,Liu Y
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Thu Aug 01 00:00:00 UTC 2019
Molecular profiling of primary uveal melanomas with tumor-infiltrating lymphocytes.
Oncoimmunology
Triozzi PL,Schoenfield L,Plesec T,Saunthararajah Y,Tubbs RR,Singh AD
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Wed Feb 03 00:00:00 UTC 2021
Immunogenomic analyses associate immunological alterations with mismatch repair defects in prostate cancer.
The Journal of clinical investigation
Nava Rodrigues D,Rescigno P,Liu D,Yuan W,Carreira S,Lambros MB,Seed G,Mateo J,Riisnaes R,Mullane S,Margolis C,Miao D,Miranda S,Dolling D,Clarke M,Bertan C,Crespo M,Boysen G,Ferreira A,Sharp A,Figueiredo I,Keliher D,Aldubayan S,Burke KP,Sumanasuriya S,Fontes MS,Bianchini D,Zafeiriou Z,Teixeira Mendes LS,Mouw K,Schweizer MT,Pritchard CC,Salipante S,Taplin ME,Beltran H,Rubin MA,Cieslik M,Robinson D,Heath E,Schultz N,Armenia J,Abida W,Scher H,Lord C,D'Andrea A,Sawyers CL,Chinnaiyan AM,Alimonti A,Nelson PS,Drake CG,Van Allen EM,de Bono JS
13-4777 was used in Immunohistochemistry-immunofluorescence to analyse the integrated immunogenomic landscape of advanced prostate cancer.
Mon Oct 01 00:00:00 UTC 2018
Increased circulating regulatory T cells in medicated people with schizophrenia.
Psychiatry research
Kelly DL,Li X,Kilday C,Feldman S,Clark S,Liu F,Buchanan RW,Tonelli LH
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Thu Nov 01 00:00:00 UTC 2018
Spontaneous Regression of Ovarian Carcinoma After Septic Peritonitis; A Unique Case Report.
Frontiers in oncology
Roelofsen T,Wefers C,Gorris MAJ,Textor JC,Massuger LFAG,de Vries IJM,van Altena AM
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Wed Oct 04 00:00:00 UTC 2023
Multicomponent analysis of the tumour microenvironment reveals low CD8 T cell number, low stromal caveolin-1 and high tenascin-C and their combination as significant prognostic markers in non-small cell lung cancer.
Oncotarget
Onion D,Isherwood M,Shridhar N,Xenophontos M,Craze ML,Day LJ,García-Márquez MA,Pineda RG,Reece-Smith AM,Saunders JH,Duffy JP,Argent RH,Grabowska AM
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Fri Jan 05 00:00:00 UTC 2018
Blocking the recruitment of naive CD4+ T cells reverses immunosuppression in breast cancer.
Cell research
Su S,Liao J,Liu J,Huang D,He C,Chen F,Yang L,Wu W,Chen J,Lin L,Zeng Y,Ouyang N,Cui X,Yao H,Su F,Huang JD,Lieberman J,Liu Q,Song E
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Sat Apr 01 00:00:00 UTC 2017
Regulatory T Cell Migration Is Dependent on Glucokinase-Mediated Glycolysis.
Immunity
Kishore M,Cheung KCP,Fu H,Bonacina F,Wang G,Coe D,Ward EJ,Colamatteo A,Jangani M,Baragetti A,Matarese G,Smith DM,Haas R,Mauro C,Wraith DC,Okkenhaug K,Catapano AL,De Rosa V,Norata GD,Marelli-Berg FM
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Tue Nov 21 00:00:00 UTC 2017
Cutting Edge: Selective Oral ROCK2 Inhibitor Reduces Clinical Scores in Patients with Psoriasis Vulgaris and Normalizes Skin Pathology via Concurrent Regulation of IL-17 and IL-10.
Journal of immunology (Baltimore, Md. : 1950)
Zanin-Zhorov A,Weiss JM,Trzeciak A,Chen W,Zhang J,Nyuydzefe MS,Arencibia C,Polimera S,Schueller O,Fuentes-Duculan J,Bonifacio KM,Kunjravia N,Cueto I,Soung J,Fleischmann RM,Kivitz A,Lebwohl M,Nunez M,Woodson J,Smith SL,West RF,Berger M,Krueger JG,Ryan JL,Waksal SD
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Radioimmune Assays
Mon May 15 00:00:00 UTC 2017
A novel method for assaying human regulatory T cell direct suppression of B cell effector function.
Journal of immunological methods
Weingartner E,Courneya JP,Keegan A,Golding A
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Wed Feb 01 00:00:00 UTC 2017
HDAC inhibition potentiates immunotherapy in triple negative breast cancer.
Oncotarget
Terranova-Barberio M,Thomas S,Ali N,Pawlowska N,Park J,Krings G,Rosenblum MD,Budillon A,Munster PN
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Tue Dec 26 00:00:00 UTC 2017
Long-lived pancreatic ductal adenocarcinoma slice cultures enable precise study of the immune microenvironment.
Oncoimmunology
Jiang X,Seo YD,Chang JH,Coveler A,Nigjeh EN,Pan S,Jalikis F,Yeung RS,Crispe IN,Pillarisetty VG
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Sat Nov 04 00:00:00 UTC 2023
Preferential accumulation of regulatory T cells with highly immunosuppressive characteristics in breast tumor microenvironment.
Oncotarget
Syed Khaja AS,Toor SM,El Salhat H,Faour I,Ul Haq N,Ali BR,Elkord E
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Tue May 16 00:00:00 UTC 2017
Alloantigen-specific regulatory T cells generated with a chimeric antigen receptor.
The Journal of clinical investigation
MacDonald KG,Hoeppli RE,Huang Q,Gillies J,Luciani DS,Orban PC,Broady R,Levings MK
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Fri Apr 01 00:00:00 UTC 2016
The immune microenvironment of breast ductal carcinoma in situ.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
Thompson E,Taube JM,Elwood H,Sharma R,Meeker A,Warzecha HN,Argani P,Cimino-Mathews A,Emens LA
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Tue Mar 01 00:00:00 UTC 2016
Altered expression of miR-92a correlates with Th17 cell frequency in patients with primary biliary cirrhosis.
International journal of molecular medicine
Liang DY,Hou YQ,Luo LJ,Ao L
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Fri Jul 01 00:00:00 UTC 2016
Differential tumor infiltration by T-cells characterizes intrinsic molecular subtypes in breast cancer.
Journal of translational medicine
Miyan M,Schmidt-Mende J,Kiessling R,Poschke I,de Boniface J
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry (Frozen)
Fri Jul 29 00:00:00 UTC 2016
Regulatory T Cell Modulation by CBP/EP300 Bromodomain Inhibition.
The Journal of biological chemistry
Ghosh S,Taylor A,Chin M,Huang HR,Conery AR,Mertz JA,Salmeron A,Dakle PJ,Mele D,Cote A,Jayaram H,Setser JW,Poy F,Hatzivassiliou G,DeAlmeida-Nagata D,Sandy P,Hatton C,Romero FA,Chiang E,Reimer T,Crawford T,Pardo E,Watson VG,Tsui V,Cochran AG,Zawadzke L,Harmange JC,Audia JE,Bryant BM,Cummings RT,Magnuson SR,Grogan JL,Bellon SF,Albrecht BK,Sims RJ,Lora JM
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Fri Jun 17 00:00:00 UTC 2016
Regulatory T Cell Modulation by CBP/EP300 Bromodomain Inhibition.
The Journal of biological chemistry
Ghosh S,Taylor A,Chin M,Huang HR,Conery AR,Mertz JA,Salmeron A,Dakle PJ,Mele D,Cote A,Jayaram H,Setser JW,Poy F,Hatzivassiliou G,DeAlmeida-Nagata D,Sandy P,Hatton C,Romero FA,Chiang E,Reimer T,Crawford T,Pardo E,Watson VG,Tsui V,Cochran AG,Zawadzke L,Harmange JC,Audia JE,Bryant BM,Cummings RT,Magnuson SR,Grogan JL,Bellon SF,Albrecht BK,Sims RJ,Lora JM
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Fri Jun 17 00:00:00 UTC 2016
Regulatory T Cell Modulation by CBP/EP300 Bromodomain Inhibition.
The Journal of biological chemistry
Ghosh S,Taylor A,Chin M,Huang HR,Conery AR,Mertz JA,Salmeron A,Dakle PJ,Mele D,Cote A,Jayaram H,Setser JW,Poy F,Hatzivassiliou G,DeAlmeida-Nagata D,Sandy P,Hatton C,Romero FA,Chiang E,Reimer T,Crawford T,Pardo E,Watson VG,Tsui V,Cochran AG,Zawadzke L,Harmange JC,Audia JE,Bryant BM,Cummings RT,Magnuson SR,Grogan JL,Bellon SF,Albrecht BK,Sims RJ,Lora JM
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Fri Jun 17 00:00:00 UTC 2016
Changes in peripheral blood lymphocytes in polycythemia vera and essential thrombocythemia patients treated with pegylated-interferon alpha and correlation with JAK2V617F allelic burden.
Experimental hematology & oncology
Kovacsovics-Bankowski M,Kelley TW,Efimova O,Kim SJ,Wilson A,Swierczek S,Prchal J
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Thu Oct 01 00:00:00 UTC 2020
Follicular regulatory T cells impair follicular T helper cells in HIV and SIV infection.
Nature communications
Miles B,Miller SM,Folkvord JM,Kimball A,Chamanian M,Meditz AL,Arends T,McCarter MD,Levy DN,Rakasz EG,Skinner PJ,Connick E
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunofluorescence
Tue Oct 20 00:00:00 UTC 2015
Follicular regulatory T cells impair follicular T helper cells in HIV and SIV infection.
Nature communications
Miles B,Miller SM,Folkvord JM,Kimball A,Chamanian M,Meditz AL,Arends T,McCarter MD,Levy DN,Rakasz EG,Skinner PJ,Connick E
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunofluorescence
Tue Oct 20 00:00:00 UTC 2015
Altered expression of autoimmune regulator in infant down syndrome thymus, a possible contributor to an autoimmune phenotype.
Journal of immunology (Baltimore, Md. : 1950)
Skogberg G,Lundberg V,Lindgren S,Gudmundsdottir J,Sandström K,Kämpe O,Annerén G,Gustafsson J,Sunnegårdh J,van der Post S,Telemo E,Berglund M,Ekwall O
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Mon Sep 01 00:00:00 UTC 2014
Tumor-derived exosomes promote tumor progression and T-cell dysfunction through the regulation of enriched exosomal microRNAs in human nasopharyngeal carcinoma.
Oncotarget
Ye SB,Li ZL,Luo DH,Huang BJ,Chen YS,Zhang XS,Cui J,Zeng YX,Li J
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Wed Jul 30 00:00:00 UTC 2014
HLA Class II tetramers reveal tissue-specific regulatory T cells that suppress T-cell responses in breast carcinoma patients.
Oncoimmunology
Schmidt HH,Ge Y,Hartmann FJ,Conrad H,Klug F,Nittel S,Bernhard H,Domschke C,Schuetz F,Sohn C,Beckhove P
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Flow Cytometry
Sat Jun 01 00:00:00 UTC 2013
IDO and regulatory T cell support are critical for cytotoxic T lymphocyte-associated Ag-4 Ig-mediated long-term solid organ allograft survival.
Journal of immunology (Baltimore, Md. : 1950)
Sucher R,Fischler K,Oberhuber R,Kronberger I,Margreiter C,Ollinger R,Schneeberger S,Fuchs D,Werner ER,Watschinger K,Zelger B,Tellides G,Pilat N,Pratschke J,Margreiter R,Wekerle T,Brandacher G
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Sun Jan 01 00:00:00 UTC 2012
Glucocorticosteroids modify Langerhans cells to produce TGF-β and expand regulatory T cells.
Journal of immunology (Baltimore, Md. : 1950)
Stary G,Klein I,Bauer W,Koszik F,Reininger B,Kohlhofer S,Gruber K,Skvara H,Jung T,Stingl G
13-4777 was used in Flow cytometry/Cell sorting to show that glucocorticosteroids endow Langerhans cells with regulatory T cell-promoting properties.
Sat Jan 01 00:00:00 UTC 2011
CD4(+) regulatory T cells in a cynomolgus macaque model of Mycobacterium tuberculosis infection.
The Journal of infectious diseases
Green AM,Mattila JT,Bigbee CL,Bongers KS,Lin PL,Flynn JL
Sun Aug 15 00:00:00 UTC 2010
Indispensable role of the Runx1-Cbfbeta transcription complex for in vivo-suppressive function of FoxP3+ regulatory T cells.
Immunity
Kitoh A,Ono M,Naoe Y,Ohkura N,Yamaguchi T,Yaguchi H,Kitabayashi I,Tsukada T,Nomura T,Miyachi Y,Taniuchi I,Sakaguchi S
13-4777 was used in Flow cytometry/Cell sorting to investigate the role of Runx1-Cbfbeta in Treg functions.
Fri Oct 16 00:00:00 UTC 2009
Gamma c-signaling cytokines induce a regulatory T cell phenotype in malignant CD4+ T lymphocytes.
Journal of immunology (Baltimore, Md. : 1950)
Kasprzycka M,Zhang Q,Witkiewicz A,Marzec M,Potoczek M,Liu X,Wang HY,Milone M,Basu S,Mauger J,Choi JK,Abrams JT,Hou JS,Rook AH,Vonderheid E,Woetmann A,Odum N,Wasik MA
Published figure using FOXP3 monoclonal antibody (Product # 13-4777-82) in Immunohistochemistry
Fri Aug 15 00:00:00 UTC 2008
Early resolution of acute immune activation and induction of PD-1 in SIV-infected sooty mangabeys distinguishes nonpathogenic from pathogenic infection in rhesus macaques.
Journal of immunology (Baltimore, Md. : 1950)
Estes JD,Gordon SN,Zeng M,Chahroudi AM,Dunham RM,Staprans SI,Reilly CS,Silvestri G,Haase AT
Thu May 15 00:00:00 UTC 2008