Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEDEGWFDP WCLLQGLRRK VQSLGVLFCQ GEVTRFVSSS QRMLTTDDKA VVLKRIHEVH VKMDRSLEYQ PVECAIVINA AGAWSAQIAA LAGVGEGPPG TLQGTKLPVE PRKRYVYVWH CPQGPGLETP LVPDTSGAYF RREGLGSNYL GGRSPTEQEE PDPANLEVDH DFFQDKVWPH LALRVPAFET LKVQSAWAGY YDYNTFDQNG VVGPHPLVVN MYFATGFSGH GLQQAPGIGR AVAEMVLKGR FQTIDLSPFL FTRFYLGEKI QENNII (212-486 aa encoded by BC013902)
This gene encodes a protein that contains a FAD-dependent oxidoreductase domain. The encoded protein is localized to the mitochondria and may function as a chaperone protein required for the function of mitochondrial complex I. Mutations in this gene are associated with mitochondrial complex I deficiency. Alternatively spliced transcript variants have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FAD-dependent oxidoreductase domain containing 1; FAD-dependent oxidoreductase domain-containing protein 1
Gene Aliases: FOXRED1; FP634; H17
UniProt ID: (Human) Q96CU9
Entrez Gene ID: (Human) 55572
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support