Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQQQPLPGP GAPTTEPTKP PYSYIALIAM AIQSSPGQRA TLSGIYRYIM GRFAFYRHNR PGWQNSIRHN LSLNECFVKV PRDDRKPGKG SYWTLDPDCH DMFEHGSFLR RRRRFTRQTG AEGTRGPAKA RRGPLRATSQ DPGVPNATTG RQCSFPPELP DPKGLSFGGL VGAMPASMCP ATTDGRPRPP MEPKEISTPK PACPGELPVA TSSSSCPAFG FPAGFSEAES FNKAPTPVLS PESGIGSSYQ CRLQALNFCM GADPGLEHLL ASAAPSPAPP TPPGSLRAPL PLPTDHKEPW VAGGFPVQGG SGYPLGLTPC LYRTPGMFFF E (1-330 aa encoded by BC013408)
The forkhead family of transcription factors belongs to the winged helix class of DNA-binding proteins. The protein encoded by this intronless gene contains a forkhead domain and is found predominantly in aorta and kidney. The function of the encoded protein is unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Forkhead box protein S1; forkhead-like 18; Forkhead-like 18 protein; forkhead-related activator 10; Forkhead-related transcription factor 10; forkhead-related transcription factor FREAC-10; FREAC-10; Transcription factor FKH-3
Gene Aliases: Fkh3; FKHL18; FOXS1; FREAC10
UniProt ID: (Human) O43638, (Mouse) Q61574
Entrez Gene ID: (Human) 2307, (Mouse) 14239, (Rat) 311547
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support