Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: SAPLISSSPV KAAREYEDPP SEEEDKIKEE PLTISELVYN PSASLLPTPV DDDEIDMLFD CPSRLELERE DTDSFEDLEA DENAFLIA
Putative tumor suppressor gene that may be implicated in the origin and progression of lung cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4.1O; 41O; band 4.1-like protein 4; Band 4.1-like protein 4O; FERM domain-containing protein 3; Ovary type protein 4.1; protein 4.1O
Gene Aliases: 4.1O; EPB41L4O; EPB41LO; FRMD3; P410
UniProt ID: (Human) A2A2Y4
Entrez Gene ID: (Human) 257019
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support