Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • FRS2 Antibodies

            Invitrogen

            FRS2 Polyclonal Antibody

            1 Reference
            View all (24) FRS2 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite FRS2 Polyclonal Antibody

            • Antibody Testing Data (1)
            Application
            FRS2 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            FRS2 Antibody (PA1-24685) in ICC/IF

            Immunofluorescence analysis FRS2 Polyclonal Antibody was performed using 70% confluent log phase LNCaP cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled FRS2 polyclonal antibody (Product PA1-24685) at 1:100 dilution in 0.1% BSA, incubated at 4 degree Celsius overnight and then with Goat anti-Rabbit IgG (H+L), Superclonal™ Recombinant Secondary Antibody, Alexa Fluor 488 (Product # A28175) at... View More
            View Product

            FRS2 Antibody in Immunocytochemistry (ICC/IF)

            Product Details

            PA1-24685

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:1,000
            View 1 publication 1 publication

            Immunocytochemistry (ICC/IF)

            1:100
            -

            Immunoprecipitation (IP)

            10-20 µg/mL
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Synthetic peptide corresponding to human FRS2 sequence, (amino acids 487-505).
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1.9 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.4, with 1% BSA

            Contains

            15mM sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2247174

            Product Specific Information

            PA1-24685 detects a band of ~92 and 95 kDa. Positive control of rat PC-12 and human HepG2 cell lysates suggested.

            Target Information

            Adapter protein that links activated FGR and NGF receptors to downstream signaling pathways. Plays an important role in the activation of MAP kinases and in the phosphorylation of PIK3R1, the regulatory subunit of phosphatidylinositol 3-kinase, in response to ligand-mediated activation of FGFR1. Modulates signaling via SHC1 by competing for a common binding site on NTRK1. [UniProt]

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: FGFR signalling adaptor; FGFR substrate 2; FGFR-signaling adaptor SNT; Fibroblast growth factor receptor substrate 2; FRS2-alpha; SNT-1; SNT2; Suc1-associated neurotrophic factor target 1

            View more View less

            Gene Aliases: 4732458E18; C330018A15Rik; FRS1A; FRS2; FRS2A; FRS2alpha; SNT; SNT-1; SNT1

            View more View less

            UniProt ID: (Human) Q8WU20, (Mouse) Q8C180

            View more View less

            Entrez Gene ID: (Human) 10818, (Mouse) 327826, (Rat) 314850

            View more View less

            Function(s)

            transmembrane receptor protein tyrosine kinase adaptor activity Ras guanyl-nucleotide exchange factor activity fibroblast growth factor receptor binding insulin receptor binding neurotrophin TRKA receptor binding protein binding 1-phosphatidylinositol-3-kinase activity phosphatase activator activity phosphatidylinositol-4,5-bisphosphate 3-kinase activity

            Process(es)

            activation of MAPK activity gastrulation with mouth forming second organ induction lens development in camera-type eye ventricular septum development transmembrane receptor protein tyrosine kinase signaling pathway neuroblast proliferation fibroblast growth factor receptor signaling pathway anterior/posterior axis specification, embryo positive regulation of signal transduction forebrain development regulation of apoptotic process optic placode formation involved in camera-type eye formation regulation of epithelial cell proliferation prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis lens fiber cell development regulation of ERK1 and ERK2 cascade MAPK cascade activation of MAPKK activity transmembrane receptor protein tyrosine phosphatase signaling pathway G-protein coupled receptor signaling pathway regulation of phosphatidylinositol 3-kinase signaling phosphatidylinositol-3-phosphate biosynthetic process positive regulation of GTPase activity phosphatidylinositol phosphorylation phosphatidylinositol-mediated signaling
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0