Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MPTNGLHQVL KIQFGLVNDT DRYLTAESFG FKVNASAPSL KRKQTWVLEP DPGQGTAVLL RSSHLGRYLS AEEDGRVACE AEQPGRDCRF LVLPQPDGRW VLRSEPHGRF FGGTEDQLSC FATAVSPAEL WTVHLAIHPQ AHLLSVSRRR YVHLCPREDE MAADGDKPWG VDALLTLIFR SRRYCLKSCD SRYLRSDGRL VWEPEPRACY TLEFKAGKLA FKDCDGHYLA PVGPAGTLKA GRNTRPGKDE LFDLEESHPQ VVLVAANHRY VSVRQGVNVS ANQDDELDHE TFLMQIDQET KKCTFYSSTG
FSCN2 acts as an actin bundling protein. May play a pivotal role in photoreceptor cell-specific events, such as disk morphogenesis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fascin homolog 2, actin-bundling protein, retinal; Fascin-2; Retinal fascin
Gene Aliases: A930022G03; Ahl8; C630046B20Rik; Fscn2
UniProt ID: (Mouse) Q32M02
Entrez Gene ID: (Rat) 303741, (Mouse) 238021
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support