Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide Sequence: MPDEDSKIDH YVLEYRRTNF EGPPRLKEDQ PWMVIEGIRQ TEYTLTGLKF DMKYMNFRVK ACNKAVAGEF SEPVTLETPA FMFRLDASTS HQNLRVDDLS VEWDAMGGKV QDIKAREKDG KGRTASPINS PARGTPSPKR MPSGRGGRDR FTAESYTVLG DTLIDGGEHY WEVRYEPDSK AFGVGVAYRS LGRFEQLGKT AASWCLHVNN WLQVSFTAKH ANKVKVLDAP VPDCLGVHCD FHQGLLSFYN ARTKQ
FSD1 encodes a centrosome associated protein that is characterized by an N-terminal coiled-coil region downstream of B-box (BBC) domain, a central fibronectin type III domain, and a C-terminal repeats in splA and RyR (SPRY) domain. The encoded protein associates with a subset of microtubules and may be involved in the stability and organization of microtubules during cytokinesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fibronectin type 3 and SPRY (spla, ryanodine) domain containing (with coiled-coil motif) 1; fibronectin type 3 and SPRY domain containing 1; Fibronectin type III and SPRY domain-containing protein 1; Microtubule-associated protein GLFND; MID1-related protein 1; midline 1-related protein 1
Gene Aliases: FSD1; GLFND; MIR1; VLP27
UniProt ID: (Human) Q9BTV5
Entrez Gene ID: (Human) 79187
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support