Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ferritin L subunit; ferritin L subunit 1; ferritin L-chain; Ferritin light chain; ferritin light chain 1; ferritin light polypeptide-like 3; ferritin, light polypeptide; fril
Gene Aliases: FTL; Ftl-1; L-ferritin; LFTD; NBIA3
UniProt ID: (Human) P02792
Entrez Gene ID: (Human) 2512, (Mouse) 14325
Molecular Function: storage protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support