Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SNESHCSLQ IKALAKIHAF VQDTTLSEPR QAEIRKECLR LWGIPDQARV APSSSDPKSK FFELIQGTEI DIFSYKPTLL TSKTLEKIRP VFDYRCMVSG SEQKFLIGLG KSQIYTWDGR QSDRWIKLDL KTELPRDTLL SVEIVHELKG EGKAQRKISA IHILDVLVLN GTDVREQHFN QRIQLAEKFV KAVSKPSRPD MNPIRVKEVY RLEEMEKIFV RLEMKIIKGS SGTPKLSYTG RDDRHFVPMG LYIVRTVNEP WTMGFSKSFK KKFFYNKKTK DSTFDLPADS IAPFHICYYG RLFWEWGDGI RVHDSQKPQD QDKLSKEDVL SFIQMHRA
S-adenosyl-L-methionine-dependent methyltransferase that mediates mRNA cap1 2'-O-ribose methylation to the 5'-cap structure of mRNAs. Methylates the ribose of the first nucleotide of a m(7)GpppG-capped mRNA and small nuclear RNA (snRNA) to produce m(7)GpppRm (cap1). Displays a preference for cap0 transcripts. Cap1 modification is linked to higher levels of translation. May be involved in the interferon response pathway. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cap methyltransferase 1; Cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1; Cap1 2'O-ribose methyltransferase 1; FtsJ methyltransferase domain containing 2; FtsJ methyltransferase domain-containing protein 2; Interferon-stimulated gene 95 kDa protein; ISG95; MTr1; S-adenosyl-L-methionine-dependent methyltransferase FTSJD2
Gene Aliases: 1300018I05Rik; AI451264; CMTR1; FTSJD2; hMTr1; KIAA0082; mKIAA0082; MTR1
UniProt ID: (Human) Q8N1G2, (Mouse) Q9DBC3
Entrez Gene ID: (Human) 23070, (Mouse) 74157
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support