Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: U-87MG, Mouse brain, Mouse kidney.
Immunogen sequence: LLKAAFICAG IVHVHSELQL SEQLLRTFGG GFELHTWSEL PHGSGLGTSS ILAGTALAAL QRAAGRVVGT EALIHAVLHL EQVLTTGGGW QDQVGGLMPG IKVGRSRAQL PLKVEVEEVT VPEGFVQKLN DHLLLVYTGK TRLARNLLQD VLRSWYARLP AVVQNAHSLV RQTEECAEGF RQGSLPLLGQ CLTSYWEQKK LMAPGCEPLT VRRMMDVLAP HVHGQSLAGA GGGGFLYLLT KEPQQKEALE AVLAKTEGLG NYSIHLVEVD TQGLSLKLLG TEASTCCPFP
The protein encoded by this gene belongs to the GHMP (galacto-, homoserine, mevalonate and phosphomevalonate) kinase family and catalyzes the phosphorylation of L-fucose to form beta-L-fucose 1-phosphate. This enzyme catalyzes the first step in the utilization of free L-fucose in glycoprotein and glycolipid synthesis. L-fucose may be important in mediating a number of cell-cell interactions such as blood group antigen recognition, inflammation, and metastatis. While several transcript variants may exist for this gene, the full-length nature of only one has been described to date.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fucokinase; L-fucose kinase; mFLJ00241 protein; UNQ5838
Gene Aliases: 1110046B12Rik; AI303278; FCSK; FUK
UniProt ID: (Human) Q8N0W3, (Mouse) Q7TMC8
Entrez Gene ID: (Human) 197258, (Mouse) 234730
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support