Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QHPASLSGT WTVYPNGRFG NQMGQYATLL ALAQLNGRRA FILPAMHAAL APVFRITLPV LAPEVDSRTP WRELQLHDWM SEEYADLRDP FLKLSGFPCS WTFFHHLREQ IRREFTLHDH LREEAQSVLG QLRLGRTGDR PRTFVGVHVR RGDYLQVMPQ RWKGVVGDSA YLRQAMDWFR ARHEAPVFVV TSNGMEWCKE NIDTSQGDVT FAGDGQEATP WKDFALLTQC NHTIMTIGTF GFWAAYLAGG DTVYLANFTL PDSEFLKIFK PEAAFLPEWV GINADLSPLW TLAKP (72-365 aa encoded by B C074732)
FUT1 is a Golgi stack membrane protein that is involved in the creation of a precursor of the H antigen, which is required for the final step in the soluble A and B antigen synthesis pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-alpha-L-fucosyltransferase; alpha (1,2) fucosyltransferase; alpha(1,2) fucosyltransferase 1; Alpha(1,2)FT 1; beta-galactoside alpha-1,2-fucosyltransferase; Blood group H alpha 2-fucosyltransferase; Fucosyltransferase 1; fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase); fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, H blood group); galactoside 2-alpha-L-fucosyltransferase 1; Galactoside alpha-(1,2)-fucosyltransferase 1; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1; H transferase; MFUT-1; Type 1 galactoside alpha-(1,2)-fucosyltransferase FUT1; Type 2 galactoside alpha-(1,2)-fucosyltransferase FUT1
Gene Aliases: FUT1; H; HH; HSC
UniProt ID: (Human) P19526, (Mouse) O09160
Entrez Gene ID: (Human) 2523, (Mouse) 14343
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support