Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The FUT11 gene encodes for fructosyltransferase 11, an enzyme that is part of the glycosyltransferase family involved in the modification of glycoproteins and glycolipids by catalyzing the addition of fucose moieties. Enzymes like FUT11 play a critical role in the biosynthesis of Lewis antigen structures, significantly contributing to cell-cell adhesion, signal transduction, and immune recognition. Fucosylation, the process mediated by fructosyltransferases, is crucial for various biological functions, including the regulation of leukocyte trafficking and selectin binding, emphasizing its importance in immune responses and inflammation. While FUT11 is less characterized compared to other fucosyltransferases, altered expression patterns and functional abnormalities in these enzymes are linked to pathological conditions, such as cancer and inflammatory diseases, where changes in glycosylation can affect tumor progression, metastasis, and immune evasion. Ongoing research into FUT11 aims to elucidate its specific role in cellular processes and explore potential therapeutic applications by targeting fucosylation pathways to modify disease progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha (1,3) fucosyltransferase; Alpha-(1,3)-fucosyltransferase 11; alpha3-fucosyltransferase 11; EC 2.4.1; Fuc-TXI; Fucosyltransferase 11; fucosyltransferase 11 (alpha (1,3) fucosyltransferase); Fucosyltransferase XI; fucT-XI; Galactoside 3-L-fucosyltransferase 11; MGC119338; MGC119339; MGC33202
Gene Aliases: 3110009G03Rik; AI317183; FUT11
UniProt ID: (Human) Q495W5, (Mouse) Q8BHC9, (Rat) Q68FV3
Entrez Gene ID: (Human) 170384, (Mouse) 73068, (Rat) 286971
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support