Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: YEAFVPADAF VHVDDFGSAR ELAAFLTGMN ESRYQRFFAW RDRLRVRLFT DWRERFCAIC DRYPHLPRSQ VYEDLEGWFQ A
May catalyze alpha-1,3 glycosidic linkages involved in the expression of sialyl Lewis X antigens.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-(1,3)-fucosyltransferase 7; Fuc-TVII; Fucosyltransferase 7; fucosyltransferase 7 (alpha (1,3) fucosyltransferase); Fucosyltransferase VII; Galactoside 3-L-fucosyltransferase; Selectin ligand synthase; selectin-ligand synthase
Gene Aliases: FucT-VII; FUT7
UniProt ID: (Human) Q11130
Entrez Gene ID: (Human) 2529
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support